Learn More
Thermco Products Inc ASTM NM THERM FC -20-150C
Catalog No. NC1204217
Encompass_Preferred
ASTM S1C thermometer (Blue spirit) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.
Catalog No. NC1204217 Supplier Thermco Products Inc Supplier No. ACC001CBLSFC
ASTM S1C thermometer (Blue spirit) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.