Learn More
Gilson Company Inc Soil Hydrometer, ASTM 152H with Certificate of Conformity
Catalog No. NC0655296
Encompass
Soil Hydrometer, ASTM 152H measures in grams per liter (g/L) of suspension and has a range of -5 — +60 g/L, in 1g/L divisions at 68°F (20°C). SA-2 hydrometer comes with a certificate of conformity.
Catalog No. NC0655296 Supplier Gilson Company Inc Supplier No. SA2
Soil Hydrometer, ASTM 152H measures in grams per liter (g/L) of suspension and has a range of -5 — +60 g/L, in 1g/L divisions at 68°F (20°C). SA-2 hydrometer comes with a certificate of conformity.