Learn More
Thermco Products Inc ASTM THERMOMETER WITH CERT
Catalog No. NC1409654
Encompass_Preferred
ASTM Thermometer with Cert S120C Kinimatic Viscosity +38.6 to 41.4°C 0.05°C TOTAL 310mm
Catalog No. NC1409654 Supplier Thermco Products Inc Supplier No. ACC120CBLS
ASTM Thermometer with Cert S120C Kinimatic Viscosity +38.6 to 41.4°C 0.05°C TOTAL 310mm