Learn More
Thermco Products Inc ASTM 0S67C Approved Precision Thermometer, 95/155C x 0.2C, Blue Spirit, Total Imm, 40mm long
Catalog No. NC0772215
Encompass_Preferred
Thermco’s ASTM - Precision Non mercury Thermometers are calibrated against NlST standards and comes with a calibration document showing corrections to the nearest tenth of a scale division traceable to NIST. Certified at one temperatur point. The thermometer meets with all ASTM specifications and has an ice point scale enabling the user to validate and update the calibration document to insure accuracy.
Catalog No. NC0772215 Supplier Thermco Products Inc Supplier No. ACC067CBLS
Thermco’s ASTM - Precision Non mercury Thermometers are calibrated against NlST standards and comes with a calibration document showing corrections to the nearest tenth of a scale division traceable to NIST. Certified at one temperatur point. The thermometer meets with all ASTM specifications and has an ice point scale enabling the user to validate and update the calibration document to insure accuracy.