Learn More
Aviva Systems Biology Corp ASTN2 Antibody - N-terminal region (ARP46963_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46963_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASTN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ASTN2. Alternate names: KIAA0634, bA67K19.1ApplicationWB, ICC. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-59446