Learn More
Cayman Chemical Astoloca mnsuns Astolon 10mg
SDP
Catalog No. 502912701
Encompass_Preferred
A sesquiterpene with anticancer activity; cytotoxic to H460, ES-2, and DU145 cells (IC50s = 51.3, 61.5, and 58 UG/ml, respectively)
Catalog No. 50-291-2701 Supplier Cayman Chemical Supplier No. 3615710 MG
A sesquiterpene with anticancer activity; cytotoxic to H460, ES-2, and DU145 cells (IC50s = 51.3, 61.5, and 58 UG/ml, respectively)