Learn More
Cayman Chemical Aristolone, Purity: ≥98%, CAS: 25274-27-5, 50 mg
SDP
Catalog No. 502912702
Encompass_Preferred
A sesquiterpene with anticancer activity; cytotoxic to H460, ES-2, and DU145 cells (IC50s = 51.3, 61.5, and 58 µg/ml, respectively)
Catalog No. 50-291-2702 Supplier Cayman Chemical Supplier No. 3615750 MG
A sesquiterpene with anticancer activity; cytotoxic to H460, ES-2, and DU145 cells (IC50s = 51.3, 61.5, and 58 µg/ml, respectively)