Learn More
Cayman Chemical AstragalosIde I 10mg
SDP
Catalog No. 502987912
Encompass_Preferred
A saponin from Astragalus; has osteogenic activities in vitro; induces differentiation and matrix mineralization of MC3T3-E1 cells; increases alkaline phosphatase activity; increases expression of osteoblast-related genes (10-40 µM), including Wnt/β-catenin
Catalog No. 50-298-7912 Supplier Cayman Chemical Supplier No. 2243110 MG
A saponin from Astragalus; has osteogenic activities in vitro; induces differentiation and matrix mineralization of MC3T3-E1 cells; increases alkaline phosphatase activity; increases expression of osteoblast-related genes (10-40 µM), including Wnt/β-catenin