Learn More
Medchemexpress LLC Astragaloside I | 84680-75-1 | 869.04 | 100 MG
SDP
Catalog No. 5000389670
Encompass_Preferred
Astragaloside I is a main active ingredient in Astragalus membranaceus that possesses osteogenic properties. It stimulates osteoblast differentiation by activating the Wnt/β-catenin signaling pathway. This product is for research use only.
  • Possesses osteogenic properties
  • Stimulates osteoblast differentiation
  • Activates the Wnt/β-catenin signaling pathway
  • Upregulates expression of osteogenesis marker genes
Catalog No. 50-003-89670 Supplier Medchemexpress LLC Supplier No. HYN0432100MG
Astragaloside I is a main active ingredient in Astragalus membranaceus that possesses osteogenic properties. It stimulates osteoblast differentiation by activating the Wnt/β-catenin signaling pathway. This product is for research use only.
  • Possesses osteogenic properties
  • Stimulates osteoblast differentiation
  • Activates the Wnt/β-catenin signaling pathway
  • Upregulates expression of osteogenesis marker genes