Learn More
Medchemexpress LLC Astragaloside II | 84676-89-1 | 827.01 | 100 MG
SDP
Catalog No. 5000389703
Encompass_Preferred
Astragaloside II is an orally active Cycloartane-type triterpene glycoside extracted from *Astragalus membranaceus*. It is used in the research of diseases such as liver cancer, osteoporosis, immunosuppressive diseases, and ulcerative colitis. It functions by inhibiting autophagy, decreasing pro-inflammatory cytokines (IL-6, IL-1β), HIF-α, p-p65, and p-IκB, and increasing superoxide dismutase (SOD). It also regulates immunity and reduces inflammatory responses.
  • Promotes cell viability of rat primary osteoblasts.
  • Enhances proliferation of primary splenocytes.
  • Sensitizes cells to 5-fluorouracil-induced cell death.
  • Inhibits levels of HIF-α, p-p65, and p-IκB, demonstrating an anti-inflammatory effect.
  • Alleviates symptoms of DSS-induced ulcerative colitis in mice.
  • Reduces DAI score and prevents body weight loss in vivo.
  • Increases colon length and superoxide dismutase in colon tissues.
Catalog No. 50-003-89703 Supplier Medchemexpress LLC Supplier No. HYN0433100MG
Astragaloside II is an orally active Cycloartane-type triterpene glycoside extracted from *Astragalus membranaceus*. It is used in the research of diseases such as liver cancer, osteoporosis, immunosuppressive diseases, and ulcerative colitis. It functions by inhibiting autophagy, decreasing pro-inflammatory cytokines (IL-6, IL-1β), HIF-α, p-p65, and p-IκB, and increasing superoxide dismutase (SOD). It also regulates immunity and reduces inflammatory responses.
  • Promotes cell viability of rat primary osteoblasts.
  • Enhances proliferation of primary splenocytes.
  • Sensitizes cells to 5-fluorouracil-induced cell death.
  • Inhibits levels of HIF-α, p-p65, and p-IκB, demonstrating an anti-inflammatory effect.
  • Alleviates symptoms of DSS-induced ulcerative colitis in mice.
  • Reduces DAI score and prevents body weight loss in vivo.
  • Increases colon length and superoxide dismutase in colon tissues.