Learn More
Cayman Chemical AstragalosIde III 10mg
SDP
Catalog No. 502986880
Encompass_Preferred
A saponin from Astragalus; bioavailable after oral administration; distributed to the thymus and spleen; has a half-life of approximately 2 hours
Catalog No. 50-298-6880 Supplier Cayman Chemical Supplier No. 2243210 MG
A saponin from Astragalus; bioavailable after oral administration; distributed to the thymus and spleen; has a half-life of approximately 2 hours