Learn More
Medchemexpress LLC Astragaloside III | 84687-42-3 | 784.97 | 1 ML
SDP
Catalog No. 5000389124
Encompass_Preferred
Astragaloside III is a triterpenoid saponin extracted from the Astragalus root. It increases NKG2D and induces TACE phosphorylation, and has been shown to induce Apoptosis. It also exhibits antiviral activity against dengue serotypes 1 and 3, and anticancer activity against breast and colon cancer.
  • Triterpenoid saponin
  • Extracted from Astragalus root
  • Increases NKG2D and induces TACE phosphorylation
  • Induces Apoptosis
  • Antiviral activity against dengue serotypes 1 and 3
  • Anticancer activity against breast and colon cancer
Catalog No. 50-003-89124 Supplier Medchemexpress LLC Supplier No. HYN043410MM/1ML
Astragaloside III is a triterpenoid saponin extracted from the Astragalus root. It increases NKG2D and induces TACE phosphorylation, and has been shown to induce Apoptosis. It also exhibits antiviral activity against dengue serotypes 1 and 3, and anticancer activity against breast and colon cancer.
  • Triterpenoid saponin
  • Extracted from Astragalus root
  • Increases NKG2D and induces TACE phosphorylation
  • Induces Apoptosis
  • Antiviral activity against dengue serotypes 1 and 3
  • Anticancer activity against breast and colon cancer