Learn More
Cayman Chemical AstragalosIde IV 1mg
SDP
Catalog No. 502955477
Encompass_Preferred
A bioactive saponin from traditional Chinese medicine; dose-dependently inhibits human adenovirus type 3 (HAdV-3) in A549 cells (IC50 = 23 µM; LC50 = 865 µM); has diverse protective effects for the cardiovascular, immune, digestive, and nervous systems; reduces myocardial infarct size in dogs
Catalog No. 50-295-5477 Supplier Cayman Chemical Supplier No. 120691 MG
A bioactive saponin from traditional Chinese medicine; dose-dependently inhibits human adenovirus type 3 (HAdV-3) in A549 cells (IC50 = 23 µM; LC50 = 865 µM); has diverse protective effects for the cardiovascular, immune, digestive, and nervous systems; reduces myocardial infarct size in dogs