Learn More
Selleck Chemical LLC Astragaloside IV S3901-25mg
SDP
Catalog No. 501948344
Encompass_Preferred
Astragaloside IV (AST-IV AS-IV) is a bioactive saponin first isolated from the dried plant roots of the genus Astragalus which is used in traditional Chinese medicine It has various effect on the cardiovascular immune digestive and nervous systems AS-IV suppresses activation of p-Akt p-mTOR p-NF- B and p-Erk1/2
Catalog No. 50-194-8344 Supplier Selleck Chemical LLC Supplier No. S390125MG
Astragaloside IV (AST-IV AS-IV) is a bioactive saponin first isolated from the dried plant roots of the genus Astragalus which is used in traditional Chinese medicine It has various effect on the cardiovascular immune digestive and nervous systems AS-IV suppresses activation of p-Akt p-mTOR p-NF- B and p-Erk1/2