Learn More
Medchemexpress LLC Astragaloside IV | 84687-43-4 | 784.97 | 1 ML
SDP
Catalog No. 5000389930
Encompass_Preferred
Astragaloside IV, an active component isolated from *Astragalus membranaceus*, suppresses the activation of ERK1/2 and JNK, and downregulates matrix metalloproteases (MMP)-2, (MMP)-9 in MDA-MB-231 breast cancer cells.
  • Suppresses ERK1/2 and JNK activation
  • Downregulates matrix metalloproteases (MMP)-2 and (MMP)-9
  • Inhibits NSCLC cell growth and increases chemosensitivity to cisplatin
  • Inhibits viability and invasive potential of MDA-MB-231 breast cancer cells
  • Prevents cognitive deficits from cerebral ischemia and reperfusion
  • Decreases cytokine levels and inhibits TLR4 pathways
  • Attenuates NLRP3 and cleaved-caspase-1 expression
  • Reduces serum ALT/AST levels and liver apoptosis
  • Increases 48-hour survival rate in mice
Catalog No. 50-003-89930 Supplier Medchemexpress LLC Supplier No. HYN043110MM/1ML
Astragaloside IV, an active component isolated from *Astragalus membranaceus*, suppresses the activation of ERK1/2 and JNK, and downregulates matrix metalloproteases (MMP)-2, (MMP)-9 in MDA-MB-231 breast cancer cells.
  • Suppresses ERK1/2 and JNK activation
  • Downregulates matrix metalloproteases (MMP)-2 and (MMP)-9
  • Inhibits NSCLC cell growth and increases chemosensitivity to cisplatin
  • Inhibits viability and invasive potential of MDA-MB-231 breast cancer cells
  • Prevents cognitive deficits from cerebral ischemia and reperfusion
  • Decreases cytokine levels and inhibits TLR4 pathways
  • Attenuates NLRP3 and cleaved-caspase-1 expression
  • Reduces serum ALT/AST levels and liver apoptosis
  • Increases 48-hour survival rate in mice