Learn More
Medchemexpress LLC HY-N6577 5mg Medchemexpress, Astragaloside VI CAS:84687-45-6 Purity:>98%
SDP
Catalog No. 501933958
Encompass_Preferred
Medchemexpress, HY-N6577 5mg Astragaloside VI CAS:84687-45-6 Astragaloside VI could activate EGFR/ERK signalling pathway to improve wound healing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3958 Supplier Medchemexpress LLC Supplier No. HYN65775MG
Medchemexpress, HY-N6577 5mg Astragaloside VI CAS:84687-45-6 Astragaloside VI could activate EGFR/ERK signalling pathway to improve wound healing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.