Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Astrazon Pink FG, TCI America™
SDP

Catalog No. A086725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A086725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A086725G Supplier TCI America Supplier No. A086725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3648-36-0
Molecular Formula C22H26Cl2N2
Molecular Weight (g/mol) 389.364
MDL Number MFCD00031747
InChI Key ZTBANYZVKCGOKD-UHFFFAOYSA-M
Synonym Basic Red 13, Stenacrile Pink G
PubChem CID 6433371
IUPAC Name N-(2-chloroethyl)-N-methyl-4-[(E)-2-(1,3,3-trimethylindol-1-ium-2-yl)ethenyl]aniline;chloride
SMILES CC1(C2=CC=CC=C2[N+](=C1C=CC3=CC=C(C=C3)N(C)CCCl)C)C.[Cl-]

Specifications

Color Purple-Red
Quantity 25 g
Formula Weight 389.36
Physical Form Crystalline Powder
Chemical Name or Material Astrazon Pink FG
EINECSNumber (5)-1950
RTECSNumber NM2248000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.