Learn More
CPC Scientific H-DPhe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Nle-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-cyclo(Glu-Ala-His-Lys)-Asn-Arg-Lys-Leu-Nle-Glu-Ile-Ile-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CRFS003B
Encompass
A new CRF antagonist (cyclo(30/33) that, when injected into the CSF at low doses (1-10ug), results in antagonistic action against CRF and stress-related alterations of gastrointestinal motor function, without an intrinsic effect in these in vivo systems.ONE-LETTER SEQUENCE: fHLLREVLE-Nle-ARAEQLAQ-cyclo(EAHK)NRKL-Nle-EII-NH2MOLECULAR FORMULA: C161H269N49O42MOLECULAR WEIGHT:3563.24STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [170809-51-5], SYNONYMS: (D-Phe,Nle,cyclo(Glu-Lys))-CRF (12-41) (human, rat)RESEARCH AREA: NeuropeptidesREFERENCES: V. Martinez et al., J. Pharmacol. Exp. Ther., 280(2), 754 (1997)
Catalog No. 50-210-9164