Learn More
Medchemexpress LLC Astringin | 29884-49-9 | 1 ML
SDP
Catalog No. 5000418307
Encompass_Preferred
Astringin is an orally active natural flavonoid compound. It inhibits the production of oxidative stress and inflammatory factors. This compound has multiple activities, including anti-oxidation, anti-inflammation, and anti-apoptosis, and is also an inhibitor of ferroptosis. Astringin can be used in the research of diseases such as acute lung injury.
  • Inhibits oxidative stress and inflammatory factors
  • Exhibits anti-oxidation, anti-inflammation, and anti-apoptosis activities
  • Functions as an inhibitor of ferroptosis
  • Suitable for research on diseases like acute lung injury
Catalog No. 50-004-18307 Supplier Medchemexpress LLC Supplier No. HYN409310MM/1ML
Astringin is an orally active natural flavonoid compound. It inhibits the production of oxidative stress and inflammatory factors. This compound has multiple activities, including anti-oxidation, anti-inflammation, and anti-apoptosis, and is also an inhibitor of ferroptosis. Astringin can be used in the research of diseases such as acute lung injury.
  • Inhibits oxidative stress and inflammatory factors
  • Exhibits anti-oxidation, anti-inflammation, and anti-apoptosis activities
  • Functions as an inhibitor of ferroptosis
  • Suitable for research on diseases like acute lung injury