Learn More
MSC Monroe Fluid Technology All-Purpose Cleaner, Low Foam, Alkaline, 1 gal
Catalog No. 502817849
Encompass
This all-purpose cleaner is a low foam alkaline solution designed to effectively remove process oils, gummy deposits, grease, swarf, and common shop soils.
  • Low foam alkaline cleaner
  • Removes process oils, gummy deposits of oil, grease, swarf, and normal shop soils
  • One gallon bottle
  • Mild scent
  • Foam consistency
  • Concentrate solution
Catalog No. 50-281-7849 Supplier MSC Supplier No. 14605109
This all-purpose cleaner is a low foam alkaline solution designed to effectively remove process oils, gummy deposits, grease, swarf, and common shop soils.
  • Low foam alkaline cleaner
  • Removes process oils, gummy deposits of oil, grease, swarf, and normal shop soils
  • One gallon bottle
  • Mild scent
  • Foam consistency
  • Concentrate solution