Learn More
MSC Monroe Fluid Technology Alkaline Parts Cleaner, Rust Preventative, 55 gal.
Catalog No. 502868069
Encompass
ASTRO-CLEAN D is a high-performance, mild alkaline liquid cleaner designed to be diluted with water for various cleaning operations. It effectively cleans a variety of soils while protecting surfaces from rust and white metal corrosion, and its ability to split out oils makes disposal safer and more convenient.
  • Mild alkaline liquid cleaner
  • Removes shop soils, coolants, and emulsifiable oils
  • Safe on all surfaces
  • Actively inhibits rust and white metal corrosion
  • Conveniently splits out oils for safe disposal
Catalog No. 50-286-8069 Supplier MSC Supplier No. 14605034
ASTRO-CLEAN D is a high-performance, mild alkaline liquid cleaner designed to be diluted with water for various cleaning operations. It effectively cleans a variety of soils while protecting surfaces from rust and white metal corrosion, and its ability to split out oils makes disposal safer and more convenient.
  • Mild alkaline liquid cleaner
  • Removes shop soils, coolants, and emulsifiable oils
  • Safe on all surfaces
  • Actively inhibits rust and white metal corrosion
  • Conveniently splits out oils for safe disposal