Learn More
MSC Monroe Fluid Technology All-Purpose Cleaner, Liquid, Alkaline, 1 gal
Catalog No. 502817855
Encompass
Monroe Fluid Technology's All-Purpose Cleaner is a liquid, mild-scented solution designed for industrial cleaning. It effectively removes tough shop soils and rubber tire marks, and can be diluted for various cleaning tasks.
  • Fast-acting, deep-penetrating liquid alkaline cleaner
  • Effective for removing rubber tire marks and tough shop soils
  • Can be used as received or diluted for lighter applications
  • Suitable for auto scrubbers, agitated cleaner tanks, or manual cleaning
  • Mild scent
  • Comes in a 1 gal bottle
Catalog No. 50-281-7855 Supplier MSC Supplier No. 14605281
Monroe Fluid Technology's All-Purpose Cleaner is a liquid, mild-scented solution designed for industrial cleaning. It effectively removes tough shop soils and rubber tire marks, and can be diluted for various cleaning tasks.
  • Fast-acting, deep-penetrating liquid alkaline cleaner
  • Effective for removing rubber tire marks and tough shop soils
  • Can be used as received or diluted for lighter applications
  • Suitable for auto scrubbers, agitated cleaner tanks, or manual cleaning
  • Mild scent
  • Comes in a 1 gal bottle