Learn More
MSC Monroe Fluid Technology ASTRO-CLEAN FLOOR, Liquid Concentrate, 5 gal
Catalog No. 502817851
Encompass
ASTRO-CLEAN FLOOR is a high-performance water-extendable liquid cleaner concentrate designed for industrial cleaning applications. It is safe for use on all surfaces and can be utilized in powered scrubbers, agitated cleaner tanks, or for manual cleaning operations.
  • High performance water extendable liquid cleaner concentrate
  • Safe on all surfaces
  • Lower disposal costs than solvent cleaners
  • Mild pleasant odor
  • Non-flammable
  • Operator friendly
  • Oils conveniently split out for safe disposal
Catalog No. 50-281-7851 Supplier MSC Supplier No. 14605208
ASTRO-CLEAN FLOOR is a high-performance water-extendable liquid cleaner concentrate designed for industrial cleaning applications. It is safe for use on all surfaces and can be utilized in powered scrubbers, agitated cleaner tanks, or for manual cleaning operations.
  • High performance water extendable liquid cleaner concentrate
  • Safe on all surfaces
  • Lower disposal costs than solvent cleaners
  • Mild pleasant odor
  • Non-flammable
  • Operator friendly
  • Oils conveniently split out for safe disposal