Learn More
Sciencell Research Laboratories Astrocyte Conditioned Medium, Serum-Free, Liquid, 100 ml, pH 7.4, Sterile filtered, Protect from light, Gel ice shipping
Catalog No. NC1094574
Encompass
ScienCell Research Laboratories Astrocyte Conditioned Medium-Serum Free (ACM-sf) is a specialized liquid medium designed for scientific research applications. This serum-free formulation is prepared from primary human astrocyte cultures, conditioned for 48 hours, and is sterile filtered. It is specifically developed to support the growth of neurons and endothelial cells in culture and has been shown to protect neurons from corticosterone-induced damage. The medium maintains a stable pH of 7.4 when equilibrated in an incubator with a 5% CO2/95% air atmosphere.
  • Supports growth of neurons and endothelial cells
  • Prepared from primary human astrocyte cultures
  • Conditioned for 48 hours
  • Sterile filtered
  • Maintains pH of 7.4
  • Volume of 100 ml
  • Stored at 4°C
  • Protect from light during storage
  • Shipped on gel ice
  • Research use only, not for clinical use
  • Country of origin: United States
Catalog No. NC1094574 Supplier Sciencell Research Laboratories Supplier No. 1811SF
ScienCell Research Laboratories Astrocyte Conditioned Medium-Serum Free (ACM-sf) is a specialized liquid medium designed for scientific research applications. This serum-free formulation is prepared from primary human astrocyte cultures, conditioned for 48 hours, and is sterile filtered. It is specifically developed to support the growth of neurons and endothelial cells in culture and has been shown to protect neurons from corticosterone-induced damage. The medium maintains a stable pH of 7.4 when equilibrated in an incubator with a 5% CO2/95% air atmosphere.
  • Supports growth of neurons and endothelial cells
  • Prepared from primary human astrocyte cultures
  • Conditioned for 48 hours
  • Sterile filtered
  • Maintains pH of 7.4
  • Volume of 100 ml
  • Stored at 4°C
  • Protect from light during storage
  • Shipped on gel ice
  • Research use only, not for clinical use
  • Country of origin: United States