Learn More
Enzo Life Sciences Astrovirus type 1 antigen (200µl)
SDP
Supplier:  Enzo Life Sciences ALX6301220200
Encompass_Preferred
Formulation: Liquid. In serum-free MEM plus Earles salts. Source: From astrovirus type 1, strain A88/2 Newcastle. Inactivated by b-propiolactone 0.1% and 4 hours at 32°C. Long Term Storage: -80°C. Applications: ELISA Positive control.
Catalog No. 50-200-9649