Learn More
Olerup Inc ASTX17.1 24 -RUO KIT
SDP
Catalog No. NC2953715
Encompass
  • Next Generation Sequencing (NGS) reagent kit for HLA typing
  • Designed for research use only (RUO)
  • Utilizes hybrid capture technology
  • Supports easy single-tube workflow
  • No long-range PCR method required
  • Improved allele balance and reduced dropout rates
  • No PCR artifacts or crossovers
  • Compatible with AlloSeq Assign software for fast data import and analysis
  • Covers 17 loci and all exons for all genes
  • Sample capacity of 24 samples
  • Optimization for Illumina sequencing systems, e.g., Illumina MiSeq
Catalog No. NC2953715 Supplier Olerup Inc Supplier No. ASTX17.1 24RUO KIT
Add to Cart
Add to Cart
Request a Quote
  • Next Generation Sequencing (NGS) reagent kit for HLA typing
  • Designed for research use only (RUO)
  • Utilizes hybrid capture technology
  • Supports easy single-tube workflow
  • No long-range PCR method required
  • Improved allele balance and reduced dropout rates
  • No PCR artifacts or crossovers
  • Compatible with AlloSeq Assign software for fast data import and analysis
  • Covers 17 loci and all exons for all genes
  • Sample capacity of 24 samples
  • Optimization for Illumina sequencing systems, e.g., Illumina MiSeq