Learn More
Selleck Chemical LLC Asunaprevir 2mg 630420-16-5 BMS-650032
SDP
Supplier:  Selleck Chemical LLC S49352MG
Encompass_Preferred
Asunaprevir (BMS-650032) is an orally bioavailable inhibitor of the hepatitis C virus enzyme serine protease NS3 that is necessary for protein processing required for viral replication. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8152