Learn More
Apexbio Technology LLC Asunaprevir (BMS-650032), 10mg. Cas: 630420-16-5 MFCD: MFCD27987900. NS3 protease inhibitor
SDP
Catalog No. NC0914095
Encompass_Preferred
630420-16-5; MW 748.29; >37.4mg/mL in DMSO; Purity = 99.22%
Catalog No. NC0914095 Supplier Apexbio Technology LLC Supplier No. A319510
630420-16-5; MW 748.29; >37.4mg/mL in DMSO; Purity = 99.22%