Learn More
IDEX Health & Science Upchurch ASY PKG ADPTR 1/4X2.1 PK 085
Catalog No. NC1734472
Encompass_Preferred
Packing Adapter, Bio-Safe 1 1/2" OD; Stainless Steel;Pressure Rating Max (psi) 6500; Pressure Rating Max (bar) 448;Diameter (in) 1 1/2; Inner Diameter (mm) 2.1
Catalog No. NC1734472 Supplier IDEX Health & Science Upchurch Supplier No. 31021ASY
Packing Adapter, Bio-Safe 1 1/2" OD; Stainless Steel;Pressure Rating Max (psi) 6500; Pressure Rating Max (bar) 448;Diameter (in) 1 1/2; Inner Diameter (mm) 2.1