Learn More
Hach Company General Purpose Contacting Conductivity Sensor, Ryton Body, K=10.0 Cell Constant
Catalog No. 501997442 Shop All Hach Company Products
Encompass
Nominal Cell Constant (K) = 10.0, 3/4 inch male NPT threaded Ryton body, 6m (20ft) Analog Cable, Graphite Electrode
Catalog No. 50-199-7442 Supplier Hach Company Supplier No. 3433E8A
Nominal Cell Constant (K) = 10.0, 3/4 inch male NPT threaded Ryton body, 6m (20ft) Analog Cable, Graphite Electrode