Learn More
Waters Corp Waters Corporation Tubing Assembly, Stainless Steel, 22.5 in., Low Pressure, Column Outlet to Detector Inlet
Catalog No. NC1511985 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is an ACQUITY welded tube assembly constructed from stainless steel and measures 22.5 in. long. It is designed for low-pressure applications, specifically for connections from column outlets to detector inlets.
  • Welded tube assembly
  • Stainless steel construction
  • 22.5 in. length
  • Designed for low-pressure applications
  • Suitable for connections from column outlets to detector inlets
  • Single unit packaging
  • Replaced as required
Catalog No. NC1511985 Supplier Waters Corp Supplier No. 700005480
This tubing assembly is an ACQUITY welded tube assembly constructed from stainless steel and measures 22.5 in. long. It is designed for low-pressure applications, specifically for connections from column outlets to detector inlets.
  • Welded tube assembly
  • Stainless steel construction
  • 22.5 in. length
  • Designed for low-pressure applications
  • Suitable for connections from column outlets to detector inlets
  • Single unit packaging
  • Replaced as required