Learn More
Medchemexpress LLC HY-112618A 100mg Medchemexpress, Thalidomide-O-amido-C6-NH2 (TFA) CAS:1950635-14-9 Purity:>98%
SDP
Catalog No. 501969119
Encompass_Preferred
Thalidomide-O-amido-C6-NH2 TFA (Cereblon Ligand-Linker Conjugates 11 TFA), a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker, can be used in the synthesis of PROTACs[1].CAS# 1950635-14-9
Catalog No. 50-196-9119 Supplier Medchemexpress LLC Supplier No. HY112618A100MG
Add to Cart
Add to Cart
Thalidomide-O-amido-C6-NH2 TFA (Cereblon Ligand-Linker Conjugates 11 TFA), a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker, can be used in the synthesis of PROTACs[1].CAS# 1950635-14-9