Learn More
Medchemexpress LLC Asymmetric-dimethylarginine-d6 dihydrochloride | 1313730-20-9 | 99.5% | 281.21 | 1 MG
SDP
Catalog No. 5000438531
Encompass_Preferred
Asymmetric-dimethylarginine-d6 (dihydrochloride) is the deuterium labeled Asymmetric-dimethylarginine dihydrochloride. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-38531 Supplier Medchemexpress LLC Supplier No. HY113216BS1MG
Asymmetric-dimethylarginine-d6 (dihydrochloride) is the deuterium labeled Asymmetric-dimethylarginine dihydrochloride. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.