Learn More
Aviva Systems Biology Corp ASZ1 Antibody - middle region (ARP52537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASZ1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ASZ1. Alternate names: ALP1, ANKL1, C7orf7, GASZ, MGC26634, Orf3, CT1.19ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-727