Learn More
Cayman Chemical Aszonapyron A 2.5mg
SDP
Catalog No. 502988744
Encompass_Preferred
A meroditerpene fungal metabolite with diverse biological activities; inhibits the growth of MCF-7, NCI H460, and A375-C5 cancer cells (GI50s = 13.6, 11.6, and 10.2 μM, respectively); active against multidrug-resistant isolates of S. aureus, E. faecalis, and E. faecium (MICs = 8, 16, and 16 μg/ml, respectively) and inhibits S. aureus biofilm formation; active against P. falciparum in vitro (IC50 = 1.34 μg/ml)
Catalog No. 50-298-8744 Supplier Cayman Chemical Supplier No. 299402.5 MG
A meroditerpene fungal metabolite with diverse biological activities; inhibits the growth of MCF-7, NCI H460, and A375-C5 cancer cells (GI50s = 13.6, 11.6, and 10.2 μM, respectively); active against multidrug-resistant isolates of S. aureus, E. faecalis, and E. faecium (MICs = 8, 16, and 16 μg/ml, respectively) and inhibits S. aureus biofilm formation; active against P. falciparum in vitro (IC50 = 1.34 μg/ml)