Learn More
Medchemexpress LLC HY-129586 5mg Medchemexpress, AT-007 CAS:2170729-29-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1295865MG
Encompass_Preferred
Medchemexpress, HY-129586 5mg AT-007 CAS:2170729-29-8 AT-007 is an orally active central nervous system (CNS) penetrant Aldose Reductase inhibitor for treatment of Galactosemia with an IC50 value of 100 pM. AT-007 reduces toxic galactitol levels and prevents disease complications in GALT deficiency rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9812