Learn More
Medchemexpress LLC AT-1002 TFA | 835872-35-0 | 99.5% | 821.91 | 1 ML
SDP
Catalog No. 5000435978
Encompass_Preferred
AT-1002 TFA, a 6-mer synthetic peptide, is a tight junction regulator and absorption enhancer. It belongs to an emerging novel class of compounds that reversibly increase paracellular transport of molecules across the epithelial barrier.
  • Can undergo Cys-Cys dimerization.
  • Treatment for up to 3 hours did not affect Caco-2 cell viability at any concentration.
  • Reduced cell viability after 24 hours at concentrations of 2.5 mg/mL and higher.
  • Cells remained viable after 24 hours if washed after 3 hours exposure, indicating it does not irreversibly damage cells.
Catalog No. 50-004-35978 Supplier Medchemexpress LLC Supplier No. HY114426A10MM/1ML
AT-1002 TFA, a 6-mer synthetic peptide, is a tight junction regulator and absorption enhancer. It belongs to an emerging novel class of compounds that reversibly increase paracellular transport of molecules across the epithelial barrier.
  • Can undergo Cys-Cys dimerization.
  • Treatment for up to 3 hours did not affect Caco-2 cell viability at any concentration.
  • Reduced cell viability after 24 hours at concentrations of 2.5 mg/mL and higher.
  • Cells remained viable after 24 hours if washed after 3 hours exposure, indicating it does not irreversibly damage cells.