Learn More
Medchemexpress LLC AT-1002 TFA | 99.5% | 821.91 | 10 MG
SDP
Catalog No. 5000437798
Encompass_Preferred
AT-1002 TFA is a 6-mer synthetic peptide that functions as a tight junction regulator and absorption enhancer. It is intended for research purposes only.
  • Reversibly increases paracellular transport of molecules across the epithelial barrier.
  • Can undergo Cys-Cys dimerization.
  • No impact on Caco-2 cell viability for up to 3 hours at concentrations of 0 to 5 mg/mL.
  • Cells remained viable after 24 hours if washed after 3 hours of exposure, indicating no irreversible damage.
Catalog No. 50-004-37798 Supplier Medchemexpress LLC Supplier No. HY114426A10MG
AT-1002 TFA is a 6-mer synthetic peptide that functions as a tight junction regulator and absorption enhancer. It is intended for research purposes only.
  • Reversibly increases paracellular transport of molecules across the epithelial barrier.
  • Can undergo Cys-Cys dimerization.
  • No impact on Caco-2 cell viability for up to 3 hours at concentrations of 0 to 5 mg/mL.
  • Cells remained viable after 24 hours if washed after 3 hours of exposure, indicating no irreversible damage.