Learn More
Selleck Chemical LLC (R)-(-)-Gossypol (AT-101) acetic acid S2812-10mM/1mL
SDP
Catalog No. 503112249
Encompass_Preferred
(R)-(-)-Gossypol (AT-101) acetic acid the R-(-) enantiomer of Gossypol acetic acid binds with Bcl-2 Bcl-xL and Mcl-1 with Ki of 0 32 M 0 48 M and 0 18 M in cell-free assays does not inhibit BIR3 domain and BID AT-101 simultaneously triggers apoptosis and a cytoprotective type of autophagy Phase 2
Catalog No. 50-311-2249 Supplier Selleck Chemical LLC Supplier No. S281210MM/1ML
(R)-(-)-Gossypol (AT-101) acetic acid the R-(-) enantiomer of Gossypol acetic acid binds with Bcl-2 Bcl-xL and Mcl-1 with Ki of 0 32 M 0 48 M and 0 18 M in cell-free assays does not inhibit BIR3 domain and BID AT-101 simultaneously triggers apoptosis and a cytoprotective type of autophagy Phase 2