Learn More
Apexbio Technology LLC AT-101, 10mg. Cas: 90141-22-3 MFCD: MFCD00017352. BH3-mimetic,gossypol enantiomer
SDP
Supplier:  Apexbio Technology LLC A319610
Encompass_Preferred
AT101, a natural product from cottonseed with a BH3-mimetic structure, was identified as a small molecule inhibitor of Bcl-2/Bcl-xL/Mcl-1 that potently induces apoptosis in various cancer cell lines [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4787