Learn More
Medchemexpress LLC AT-533 | 908112-37-8 | 98.38% | 410.51 | 5 MG
SDP
Catalog No. 5000360904
Encompass_Preferred
AT-533 is a potent Hsp90 and HSV inhibitor. It suppresses tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway. Additionally, AT-533 inhibits the activation of downstream pathways such as Akt/mTOR/p70S6K, Erk1/2, and FAK.
  • Inhibits tube formation, cell migration, and invasion of human umbilical vein endothelial cells (HUVECs).
  • Decreases TNF-α, IL-1β, and IL-6 production in RAW264.7 and BV2 cells induced by HSV-1.
  • Shows anti-angiogenic ability in a chorioallantoic membrane (CAM) model.
  • Suppresses the expression of HIF-1α/VEGF signaling pathway-related proteins in MDA-MB-231 breast cancer xenografts in mice.
  • Subacute toxicity tests showed no mortality, loss of appetite, body weight changes, or adverse reactions in Sprague-Dawley rats.
Catalog No. 50-003-60904 Supplier Medchemexpress LLC Supplier No. HY1488775MG
AT-533 is a potent Hsp90 and HSV inhibitor. It suppresses tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway. Additionally, AT-533 inhibits the activation of downstream pathways such as Akt/mTOR/p70S6K, Erk1/2, and FAK.
  • Inhibits tube formation, cell migration, and invasion of human umbilical vein endothelial cells (HUVECs).
  • Decreases TNF-α, IL-1β, and IL-6 production in RAW264.7 and BV2 cells induced by HSV-1.
  • Shows anti-angiogenic ability in a chorioallantoic membrane (CAM) model.
  • Suppresses the expression of HIF-1α/VEGF signaling pathway-related proteins in MDA-MB-231 breast cancer xenografts in mice.
  • Subacute toxicity tests showed no mortality, loss of appetite, body weight changes, or adverse reactions in Sprague-Dawley rats.