Learn More
Medchemexpress LLC AT-533 | 908112-37-8 | 98.4% | 410.51 | 1 ML
SDP
Catalog No. 5000359853
Encompass_Preferred
AT-533 is a potent inhibitor of Hsp90 and HSV, suppressing tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway. It also inhibits the activation of downstream pathways like Akt/mTOR/p70S6K, Erk1/2, and FAK, and hinders tube formation, cell migration, and invasion in human umbilical vein endothelial cells (HUVECs).
  • Potent Hsp90 and HSV inhibitor
  • Suppresses tumor growth and angiogenesis
  • Blocks HIF-1α/VEGF/VEGFR-2 signaling pathway
  • Inhibits Akt/mTOR/p70S6K, Erk1/2, and FAK pathways
  • Inhibits tube formation, cell migration, and invasion in HUVECs
  • Decreases inflammatory cytokine production in HSV-1 induced cells
  • Shows anti-angiogenic ability
Catalog No. 50-003-59853 Supplier Medchemexpress LLC Supplier No. HY14887710MM/1ML
AT-533 is a potent inhibitor of Hsp90 and HSV, suppressing tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway. It also inhibits the activation of downstream pathways like Akt/mTOR/p70S6K, Erk1/2, and FAK, and hinders tube formation, cell migration, and invasion in human umbilical vein endothelial cells (HUVECs).
  • Potent Hsp90 and HSV inhibitor
  • Suppresses tumor growth and angiogenesis
  • Blocks HIF-1α/VEGF/VEGFR-2 signaling pathway
  • Inhibits Akt/mTOR/p70S6K, Erk1/2, and FAK pathways
  • Inhibits tube formation, cell migration, and invasion in HUVECs
  • Decreases inflammatory cytokine production in HSV-1 induced cells
  • Shows anti-angiogenic ability