Learn More
Medchemexpress LLC AT-533 | 908112-37-8 | 98.4% | 410.51 | 25 MG
SDP
Catalog No. 5000363307
Encompass_Preferred
AT-533 is a potent inhibitor of Hsp90 and HSV. It inhibits tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway. Additionally, AT-533 suppresses the activation of downstream pathways such as Akt/mTOR/p70S6K, Erk1/2, and FAK. It also inhibits the tube formation, cell migration, and invasion of human umbilical vein endothelial cells (HUVECs).
  • Potent Hsp90 and HSV inhibitor.
  • Suppresses tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway.
  • Inhibits the activation of downstream pathways, including Akt/mTOR/p70S6K, Erk1/2, and FAK.
  • Inhibits the tube formation, cell migration, and invasion of human umbilical vein endothelial cells (HUVECs).
Catalog No. 50-003-63307 Supplier Medchemexpress LLC Supplier No. 25MGHY148877
AT-533 is a potent inhibitor of Hsp90 and HSV. It inhibits tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway. Additionally, AT-533 suppresses the activation of downstream pathways such as Akt/mTOR/p70S6K, Erk1/2, and FAK. It also inhibits the tube formation, cell migration, and invasion of human umbilical vein endothelial cells (HUVECs).
  • Potent Hsp90 and HSV inhibitor.
  • Suppresses tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway.
  • Inhibits the activation of downstream pathways, including Akt/mTOR/p70S6K, Erk1/2, and FAK.
  • Inhibits the tube formation, cell migration, and invasion of human umbilical vein endothelial cells (HUVECs).