Learn More
Hach Company AT1000 Bundle-Alkalinity-PT
Catalog No. 5000207838 Shop All Hach Company Products
Encompass
AP0001 AT1102 - pH/Alkalinity Application PackKit includes 1 ea AT1102 97 - Titralab AT1000-1 syringe1 ea 20253 - Sulfuric Acid-0 100N 1000mL1 ea S11M002 - pH Standard/pH 4 005 IUPAC-500mL1 ea S11M004 - pH Standard/pH 7 00 IUPAC-500mL1 ea S11M007 - Buffer-pH 10 012 IUPAC-500mL
Catalog No. 50-002-07838 Supplier Hach Company Supplier No. 8606700
Add to Cart
Add to Cart
Request a Quote
AP0001 AT1102 - pH/Alkalinity Application PackKit includes 1 ea AT1102 97 - Titralab AT1000-1 syringe1 ea 20253 - Sulfuric Acid-0 100N 1000mL1 ea S11M002 - pH Standard/pH 4 005 IUPAC-500mL1 ea S11M004 - pH Standard/pH 7 00 IUPAC-500mL1 ea S11M007 - Buffer-pH 10 012 IUPAC-500mL