Learn More
Hach Company AT1000 Bundle Chlorine Dioxide
Catalog No. 5000207433 Shop All Hach Company Products
Encompass
AP0007 AT1122 - Free Total Chlorine-Chlorine Dioxide-Sulfite Application PackKit includes 1 ea AT1122 97 - Titralab AT1000-1 syringe-2 pumps1 ea 199953 - PAO Standard Solution-0 00564N 1000mL1 ea 2155353 - Buffer Solution-Phosphate pH 7 0-1L1 ea 2834100 - KI Bulk Dispenser1 ea 141832 - Hydrochloric Acid-2 5N 100mL MDB1 ea 2333353 - Iodine Standard Solution-0 0282N-1000mL1 ea 1426810 - Chlorine-50-75MG/L PK/16 mL
Catalog No. 50-002-07433 Supplier Hach Company Supplier No. 8606600
Add to Cart
Add to Cart
Request a Quote
AP0007 AT1122 - Free Total Chlorine-Chlorine Dioxide-Sulfite Application PackKit includes 1 ea AT1122 97 - Titralab AT1000-1 syringe-2 pumps1 ea 199953 - PAO Standard Solution-0 00564N 1000mL1 ea 2155353 - Buffer Solution-Phosphate pH 7 0-1L1 ea 2834100 - KI Bulk Dispenser1 ea 141832 - Hydrochloric Acid-2 5N 100mL MDB1 ea 2333353 - Iodine Standard Solution-0 0282N-1000mL1 ea 1426810 - Chlorine-50-75MG/L PK/16 mL