Learn More
Hach Company AT1000 Bundle-Total Chlorine
Catalog No. 5000208100 Shop All Hach Company Products
Encompass
AP0007 AT1122 - Free Total Chlorine-Chlorine Dioxide-Sulfite Application PackKit includes 1 ea AT1122 97 - Titralab AT1000-1 syringe-2 pumps1 ea 199953 - PAO Standard Solution-0 00564N 1000mL1 ea 1490953 - Acetate Buffer Solution-pH 4 0-1L1 ea 2834100 - KI Bulk Dispenser1 ea 2333353 - Iodine Standard Solution-0 0282N-1000mL1 ea 1426810 - Chlorine-50-75MG/L PK/16 mL
Catalog No. 50-002-08100 Supplier Hach Company Supplier No. 8606400
Add to Cart
Add to Cart
Request a Quote
AP0007 AT1122 - Free Total Chlorine-Chlorine Dioxide-Sulfite Application PackKit includes 1 ea AT1122 97 - Titralab AT1000-1 syringe-2 pumps1 ea 199953 - PAO Standard Solution-0 00564N 1000mL1 ea 1490953 - Acetate Buffer Solution-pH 4 0-1L1 ea 2834100 - KI Bulk Dispenser1 ea 2333353 - Iodine Standard Solution-0 0282N-1000mL1 ea 1426810 - Chlorine-50-75MG/L PK/16 mL