Learn More
Hach Company AT1000 Bundle-Total Hardness
Supplier:  Hach Company 8606900
Encompass
AP0003 AT1112 - Ca Mg Hardness (ISE) Application PackKit includes 1 ea AT1112 97 - Titralab AT1000-1 syringe-1 Pump1 ea 20553 - EDTA Standard Solution-0 0200N 1000mL1 ea 2382710 - Acetylacetone-100mL1 ea 607726 - Tris-Hydroxy Methyl-Amino-ACS 1001 ea 12014 - Calcium Carbonate-PRI Standard 113G1 ea 2833449 - LR Hardness QC Standard-500mL
Catalog No. 50-002-07432
Add to Cart
Request a Quote