Learn More
Selleck Chemical LLC (R)-(-)-Gossypol acetic acid 10mg 866541-93-7 AT-101 acetic acid, (-)-Gossypol acetic acid, (R)-Gossypol acetic acid
SDP
Supplier:  Selleck Chemical LLC S281210MG
Encompass_Preferred
(R)-(-)-Gossypol (AT-101) acetic acid, the R-(-) enantiomer of Gossypol acetic acid, binds with Bcl-2, Bcl-xL and Mcl-1 with Ki of 0.32 ?M, 0.48 ?M and 0.18 ?M in cell-free assays; does not inhibit BIR3 domain and BID. AT-101 simultaneously triggers apoptosis and a cytoprotective type of autophagy. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1361973