Learn More
Medchemexpress LLC HY-100113 10mg Medchemexpress, AT2 receptor agonist C21 CAS:477775-14-7 Purity:>98%
SDP
Catalog No. 502028608
Encompass_Preferred
Medchemexpress, HY-100113 10mg AT2 receptor agonist C21 CAS:477775-14-7 AT2 receptor agonist C21 is a druglike selective angiotensin II AT2 receptor agonist with Ki values of 0.4 nM and >10 µM for the AT2 receptor and AT1 receptor, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8608 Supplier Medchemexpress LLC Supplier No. HY10011310MG
Medchemexpress, HY-100113 10mg AT2 receptor agonist C21 CAS:477775-14-7 AT2 receptor agonist C21 is a druglike selective angiotensin II AT2 receptor agonist with Ki values of 0.4 nM and >10 µM for the AT2 receptor and AT1 receptor, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.