Learn More
Medchemexpress LLC HY-50940 10mg Medchemexpress, AT7519 CAS:844442-38-2 Purity:>98%
SDP
Catalog No. 502029357
Encompass_Preferred
Medchemexpress, HY-50940 10mg AT7519 CAS:844442-38-2 AT7519 as a potent inhibitor of CDKs, with IC50s of 210, 47, 100, 13, 170, and <10 nM for CDK1, CDK2, CDK4 to CDK6, and CDK9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9357 Supplier Medchemexpress LLC Supplier No. HY5094010MG
Medchemexpress, HY-50940 10mg AT7519 CAS:844442-38-2 AT7519 as a potent inhibitor of CDKs, with IC50s of 210, 47, 100, 13, 170, and <10 nM for CDK1, CDK2, CDK4 to CDK6, and CDK9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.